Learn More
Ace Glass, Inc. Bottle only, pressure, 325mL, #25, no plug
SDP
Catalog No. 509558522
Encompass_Preferred
325mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.
Catalog No. 50-955-8522 Supplier Ace Glass, Inc. Supplier No. 8648140
325mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.