Learn More
Ace Glass, Inc. Bottle only, pressure, 950mL, #25, no plug
SDP
Catalog No. 509558523
Encompass_Preferred
950mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.
Catalog No. 50-955-8523 Supplier Ace Glass, Inc. Supplier No. 8648155
950mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.