Learn More
Chemglass Life Sciences Chem-Thread, Size 25, 32mm Tube OD, Accepts 22-25mm OD Tubing, #317 O-ring
SDP
Catalog No. CG35005
Encompass_Preferred
Chem-Thread, Size 25, 32mm Tube OD, Accepts 22-25mm OD Tubing, #317 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.
Catalog No. CG35005 Supplier Chemglass Life Sciences Supplier No. CG35005
Chem-Thread, Size 25, 32mm Tube OD, Accepts 22-25mm OD Tubing, #317 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.