Learn More
Chemglass Life Sciences #25 Internal Bushing, Open Top, 26mm Opening, PTFE, #212 Viton O-Ring
SDP
Catalog No. CG36205
Encompass_Preferred
#25 Internal Bushing, Open Top, 26mm Opening, PTFE, #212 Viton O-Ring. Bushing for use with CG-360 internal glass threads. Supplied with one Viton o-ring.
Catalog No. CG36205 Supplier Chemglass Life Sciences Supplier No. CG36205
#25 Internal Bushing, Open Top, 26mm Opening, PTFE, #212 Viton O-Ring. Bushing for use with CG-360 internal glass threads. Supplied with one Viton o-ring.