Learn More
Medchemexpress LLC NO2A-butyne | 2089035-56-1 | 98.0% | 354.40 g·mol⁻¹ | C16H26N4O5 | 50mg
SDP
Catalog No. 5000270317
Encompass_Preferred
NO2A-Butyneis a bifunctional chelator (Bifunctional Chelator BFC) and a macrocyclic NOTA derivative used for tumor pre-targeting NO2A-Butyne can be used for conjugation of peptides and radionuclides
Catalog No. 50-002-70317 Supplier Medchemexpress LLC Supplier No. HYW74962750MG
NO2A-Butyneis a bifunctional chelator (Bifunctional Chelator BFC) and a macrocyclic NOTA derivative used for tumor pre-targeting NO2A-Butyne can be used for conjugation of peptides and radionuclides