Learn More
Roboz Surgical NO3 DUMONT STAINLESS STEEL
SDP
Catalog No. NC9504594
Encompass
Dumont Tweezers; Pattern #3; .08 X .04mm Tip Size; Stainless Steel; 120mm Length; Material: Inox; Biologie Tips
Catalog No. NC9504594 Supplier Roboz Surgical Supplier No. RS4903
Dumont Tweezers; Pattern #3; .08 X .04mm Tip Size; Stainless Steel; 120mm Length; Material: Inox; Biologie Tips