Learn More
Brookfield Engineer Labs Brookfield Ametek RVHV-3 Replacement Viscometer Spindle
Catalog No. NC0837445
Encompass
Min Viscosity Range (cP)100, Max Viscosity Range (cP)1600000, Replacement Viscometer Spindle
Catalog No. NC0837445 Supplier Brookfield Engineer Labs Supplier No. RVHV3
Min Viscosity Range (cP)100, Max Viscosity Range (cP)1600000, Replacement Viscometer Spindle