Learn More
Ace Glass, Inc. #36 Ace-Thred solid PTFE plug, 7855-772 FETFE O-ring front seal
SDP
Catalog No. NC0021448
Encompass_Preferred
#36 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -125 FETFE O-ring.
Catalog No. NC0021448 Supplier Ace Glass, Inc. Supplier No. 584651
#36 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -125 FETFE O-ring.