Learn More
Chemglass Life Sciences Size #4 Metal Retaining Clip for Glass Stopcock Plugs
SDP
Catalog No. NC2747057
Encompass_Preferred
Metal retaining clip designed to secure glass stopcock plugs in size #4 stopcocks. It holds the glass plug in place to reduce displacement during handling and operation, helping maintain a secure seal in glassware assemblies.
  • Secures glass stopcock plugs in size #4 stopcocks.
  • Durable metal construction.
  • Easy to install and remove.
  • Suitable for laboratory glassware and stopcock assemblies.
Catalog No. NC2747057 Supplier Chemglass Life Sciences Supplier No. CG42314
Metal retaining clip designed to secure glass stopcock plugs in size #4 stopcocks. It holds the glass plug in place to reduce displacement during handling and operation, helping maintain a secure seal in glassware assemblies.
  • Secures glass stopcock plugs in size #4 stopcocks.
  • Durable metal construction.
  • Easy to install and remove.
  • Suitable for laboratory glassware and stopcock assemblies.