Learn More
Chemglass Life Sciences Lower Vessel Only, #40 O-Ring Joint, Airfree, Schlenk
SDP
Catalog No. AF055604
Encompass_Preferred
Lower Vessel Only, #40 O-Ring Joint, Airfree, Schlenk. Lower vessel replacement part for item AF055601. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. The complete item AF055601 is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.
Catalog No. AF055604 Supplier Chemglass Life Sciences Supplier No. AF055604
Lower Vessel Only, #40 O-Ring Joint, Airfree, Schlenk. Lower vessel replacement part for item AF055601. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. The complete item AF055601 is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.