Learn More
Chemglass Life Sciences Top Adapter Only, #40 O-Ring Joint, Airfree, Schlenk
SDP
Catalog No. AF055603
Encompass_Preferred
Top Adapter Only, #40 O-Ring Joint, Airfree, Schlenk. Top adapter replacement part for item AF055601. Top adapter has a 0-4mm Chem-Vac™ Chem-Cap valve. The complete item AF055601 is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.
Catalog No. AF055603 Supplier Chemglass Life Sciences Supplier No. AF055603
Top Adapter Only, #40 O-Ring Joint, Airfree, Schlenk. Top adapter replacement part for item AF055601. Top adapter has a 0-4mm Chem-Vac™ Chem-Cap valve. The complete item AF055601 is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.