Learn More
Chemglass Life Sciences GL-45 Cap with 34mm Aperture
SDP
Catalog No. CG19505
Encompass_Preferred
GL-45 Cap with 34mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C.
Catalog No. CG19505 Supplier Chemglass Life Sciences Supplier No. CG19505
GL-45 Cap with 34mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C.