Learn More
Chemglass Life Sciences Loosening Ring, Rodaviss, Size 45
SDP
Catalog No. CG18406
Encompass_Preferred
Loosening Ring, Rodaviss, Size 45. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.
Catalog No. CG18406 Supplier Chemglass Life Sciences Supplier No. CG18406
Loosening Ring, Rodaviss, Size 45. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.