Learn More
Chemglass Life Sciences Solid Cap, PTFE Seal, GL-45 Thread
SDP
Catalog No. CG19605
Encompass_Preferred
Solid Cap, PTFE Seal, GL-45 Thread. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C. PTFE seal is also available separately.
Catalog No. CG19605 Supplier Chemglass Life Sciences Supplier No. CG19605
Solid Cap, PTFE Seal, GL-45 Thread. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C. PTFE seal is also available separately.