Learn More
Roboz Surgical NO5 DUMONT DUMOSTAR
SDP
Catalog No. NC9387672
Encompass
Dumont Tweezers; Pattern #5; .05 X .01mm Tip Size; 110mm Length; Material: Dumostar; Biologie Tips
Catalog No. NC9387672 Supplier Roboz Surgical Supplier No. RS4978
Dumont Tweezers; Pattern #5; .05 X .01mm Tip Size; 110mm Length; Material: Dumostar; Biologie Tips