Learn More
Chemglass Life Sciences O-Ring Joint, 5mm ID, 8mm Tube OD , O-Ring Size: 110, Clamp Size: 12
SDP
Catalog No. CG12401
Encompass_Preferred
O-Ring Joint, 5mm ID, 8mm Tube OD , O-Ring Size: 110, Clamp Size: 12. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. CG12401 Supplier Chemglass Life Sciences Supplier No. CG12401
O-Ring Joint, 5mm ID, 8mm Tube OD , O-Ring Size: 110, Clamp Size: 12. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.