Learn More
Chemglass Life Sciences #50 Internal Bushing, Solid, PTFE, #229 Viton O-Ring
SDP
Supplier:  Chemglass Life Sciences CG36306
Encompass_Preferred
#50 Internal Bushing, Solid, PTFE, #229 Viton O-Ring. Solid bushing for use with CG-360 internal glass threads. Supplied with one Viton o-ring.
Catalog No. CG36306