Learn More
Chemglass Life Sciences O-Ring Joint, 50mm ID, 56mm Tube OD , O-Ring Size: 229, Clamp Size: 75
SDP
Catalog No. CG12409
Encompass_Preferred
O-Ring Joint, 50mm ID, 56mm Tube OD , O-Ring Size: 229, Clamp Size: 75. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. CG12409 Supplier Chemglass Life Sciences Supplier No. CG12409
O-Ring Joint, 50mm ID, 56mm Tube OD , O-Ring Size: 229, Clamp Size: 75. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.