Learn More
Chemglass Life Sciences Lower Vessel Only, #50 O-Ring Joint, Airfree, Schlenk
SDP
Catalog No. AF055606
Encompass_Preferred
Lower Vessel Only, #50 O-Ring Joint, Airfree, Schlenk. Top adapter replacement part for item AF055602. Top adapter has a 0-4mm Chem-Vac™ Chem-Cap valve. The complete item AF055602 is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.
Catalog No. AF055606 Supplier Chemglass Life Sciences Supplier No. AF055606
Lower Vessel Only, #50 O-Ring Joint, Airfree, Schlenk. Top adapter replacement part for item AF055602. Top adapter has a 0-4mm Chem-Vac™ Chem-Cap valve. The complete item AF055602 is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.