Learn More
Roboz Surgical NO55 DUMONT DUMOXEL
SDP
Catalog No. NC9782269
Encompass
Dumont Tweezers; Pattern #55; .05 X .01mm Tip Size; Light Shanks, Superfine Points; 110mm Length; Material: Dumoxel; Biologie Tips
Catalog No. NC9782269 Supplier Roboz Surgical Supplier No. RS5063
Dumont Tweezers; Pattern #55; .05 X .01mm Tip Size; Light Shanks, Superfine Points; 110mm Length; Material: Dumoxel; Biologie Tips