Learn More
Roboz Surgical NO55 DUMONT INOX
SDP
Catalog No. NC9424569
Encompass
Dumont Tweezers; Pattern #55; .05 X .01mm Tip Size; Light Shanks, Thin tips; 110mm Length; Material: Inox; Biologie Tips
Catalog No. NC9424569 Supplier Roboz Surgical Supplier No. RS5010
Dumont Tweezers; Pattern #55; .05 X .01mm Tip Size; Light Shanks, Thin tips; 110mm Length; Material: Inox; Biologie Tips