Learn More
Chemglass Life Sciences Clamp, Horseshoe Style, #65
SDP
Catalog No. CG15106
Encompass_Preferred
Clamp, Horseshoe Style, #65. Horseshoe shaped corrosion-resistant steel clamp for use with spherical ball and socket joints. Clamp has two machine bolts and thumbscrews 180º apart to apply uniform tension. NOTE: NOT FOR USE WITH CG-124 O-RING JOINTS.
Catalog No. CG15106 Supplier Chemglass Life Sciences Supplier No. CG15106
Clamp, Horseshoe Style, #65. Horseshoe shaped corrosion-resistant steel clamp for use with spherical ball and socket joints. Clamp has two machine bolts and thumbscrews 180º apart to apply uniform tension. NOTE: NOT FOR USE WITH CG-124 O-RING JOINTS.