Learn More
Ace Glass, Inc. Connector, Ace-Safe, #7 to 1/4inid tubing
SDP
Supplier:  Ace Glass, Inc. 585306
Encompass_Preferred
Ace-Safe polypropylene tubing connector, used to connect 1/4 I.D. flexible tubing to #7 Ace-Threds, for easy, safe connect/disconnect. Includes silicone O-ring and nylon bushing which are used to make a compression seal. Maximum temperature is 110 degrees C.
Catalog No. 50-955-7178