Learn More
Roboz Surgical NO7 DUMONT STAINLESS STEEL
SDP
Catalog No. NC9683267
Encompass
Dumont Tweezers; Pattern #7; .07 X .03mm Tip Size; 115mm Length; Material: Inox; Biologie Tips
Catalog No. NC9683267 Supplier Roboz Surgical Supplier No. RS4907
Dumont Tweezers; Pattern #7; .07 X .03mm Tip Size; 115mm Length; Material: Inox; Biologie Tips