Learn More
Ace Glass, Inc. NO7 GLASS MEMBER (GB)NO7 GLASS
SDP
Catalog No. 509556895
Encompass_Preferred
12.7mm O.D. medium wall mini straight tube adapter with #7 Ace-Thred at one end for use with #7 Nylon bushing. Length is approximately 114 mm.
Catalog No. 50-955-6895 Supplier Ace Glass, Inc. Supplier No. 502705
12.7mm O.D. medium wall mini straight tube adapter with #7 Ace-Thred at one end for use with #7 Nylon bushing. Length is approximately 114 mm.