Learn More
Chemglass Life Sciences O-Ring Joint, 75mm ID, 80mm Tube OD , O-Ring Size: 341, Clamp Size: 102
SDP
Catalog No. CG12410
Encompass_Preferred
O-Ring Joint, 75mm ID, 80mm Tube OD , O-Ring Size: 341, Clamp Size: 102. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. CG12410 Supplier Chemglass Life Sciences Supplier No. CG12410
O-Ring Joint, 75mm ID, 80mm Tube OD , O-Ring Size: 341, Clamp Size: 102. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.