Learn More
Chemglass Life Sciences Adapter, #9 O-Ring to 9mm Tubulation, Airfree, Schlenk
SDP
Catalog No. AF045108
Encompass_Preferred
Replacement parts for AF-0451-01. #9 o-ring joint to 3/8" tubulation for Cajon adapters.
Catalog No. AF045108 Supplier Chemglass Life Sciences Supplier No. AF045108
Replacement parts for AF-0451-01. #9 o-ring joint to 3/8" tubulation for Cajon adapters.