Learn More
Chemglass Life Sciences Stopper #9, Pennyhead, Flask Length, Solid
SDP
Catalog No. CG301801
Encompass_Preferred
Stopper #9, Pennyhead, Flask Length, Solid. All sizes are solid with the exception of the #38 (CG-3018-08) which is hollow.
Catalog No. CG301801 Supplier Chemglass Life Sciences Supplier No. CG301801
Stopper #9, Pennyhead, Flask Length, Solid. All sizes are solid with the exception of the #38 (CG-3018-08) which is hollow.