Learn More
Enzo Life Sciences NOB (250 mg). CAS: 55894-52-5
SDP
Supplier:  Enzo Life Sciences BMLST5060250
Encompass_Preferred
Substrate for phospholipases from snake venoms and human serum. Alternative name: 4-Nitro-3-(octanoyloxy)benzoic acid. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-2192