Learn More
Aviva Systems Biology Corp NOB1 Antibody - N-terminal region (ARP54975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NOB1. Alternate names: ART-4, MST158, MSTP158, NOB1P, PSMD8BP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61383