Learn More
Selleck Chemical LLC Nobiletin S2333-5mg
SDP
Catalog No. 503112795
Encompass_Preferred
Nobiletin a citrus flavonoid isolated from citrus peels like in tangerine which has anti-inflammatory and anti-tumor activities
Catalog No. 50-311-2795 Supplier Selleck Chemical LLC Supplier No. S23335MG
Nobiletin a citrus flavonoid isolated from citrus peels like in tangerine which has anti-inflammatory and anti-tumor activities