Learn More
TARGETMOL CHEMICALS INC NOBILETIN 25MG
SDP
Catalog No. 502577041
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nobiletin (Hexamethoxyflavone) is a natural polymethoxylated flavone isolated from citrus peels with anti-inflammatory and anti-tumor activities. Nobiletin is practically insoluble (in water) and an extremely weak acidic compound (based on its pKa). purity: 98%
Catalog No. 50-257-7041 Supplier TARGETMOL CHEMICALS INC Supplier No. T283425MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nobiletin (Hexamethoxyflavone) is a natural polymethoxylated flavone isolated from citrus peels with anti-inflammatory and anti-tumor activities. Nobiletin is practically insoluble (in water) and an extremely weak acidic compound (based on its pKa). purity: 98%