Learn More
Medchemexpress LLC Nobiletin | 478-01-3 | 99.8% | 402.39 | 1 ML
SDP
Catalog No. 5000427888
Encompass_Preferred
Nobiletin is a poly-methoxylated flavone derived from citrus peel, recognized for its ability to improve memory loss. It functions as a retinoid acid receptor-related orphan receptors (RORs) agonist. This compound has demonstrated potential in decreasing reactive oxygen species (ROS) levels in differentiated C2C12 myotubes, alongside exhibiting anti-inflammatory and anti-cancer properties such as anti-angiogenesis, anti-proliferation, anti-metastasis, and apoptosis induction.
  • Improves memory loss
  • Acts as a retinoid acid receptor-related orphan receptors (RORs) agonist
  • Decreases reactive oxygen species (ROS) levels
  • Possesses anti-inflammatory properties
  • Exhibits anti-cancer properties (anti-angiogenesis, anti-proliferation, anti-metastasis)
  • Induces apoptosis
Catalog No. 50-004-27888 Supplier Medchemexpress LLC Supplier No. HYN015510MM/1ML
Nobiletin is a poly-methoxylated flavone derived from citrus peel, recognized for its ability to improve memory loss. It functions as a retinoid acid receptor-related orphan receptors (RORs) agonist. This compound has demonstrated potential in decreasing reactive oxygen species (ROS) levels in differentiated C2C12 myotubes, alongside exhibiting anti-inflammatory and anti-cancer properties such as anti-angiogenesis, anti-proliferation, anti-metastasis, and apoptosis induction.
  • Improves memory loss
  • Acts as a retinoid acid receptor-related orphan receptors (RORs) agonist
  • Decreases reactive oxygen species (ROS) levels
  • Possesses anti-inflammatory properties
  • Exhibits anti-cancer properties (anti-angiogenesis, anti-proliferation, anti-metastasis)
  • Induces apoptosis