Learn More
Medchemexpress LLC Nobiletin | 478-01-3 | MFCD03273560 | >98.0% | 402.39 | C21H22O8 | 10 MG
SDP
Catalog No. 5000296349
Encompass_Preferred
Nobiletin is a poly-methoxylated flavone derived from citrus peel. It acts as a retinoid acid receptor-related orphan receptor (ROR) agonist, modulates reactive oxygen species (ROS), and has reported antioxidant, anti-inflammatory, neuroprotective, and anti-cancer activities. Supplied as a powdered research compound; follow storage guidelines in the product datasheet.
  • Poly-methoxylated flavone with ROR agonist activity.
  • Modulates ROS and influences apoptosis and autophagy pathways.
  • Suitable for in vitro and preclinical research applications.
  • Supplied as a powder; available as a 10 MG research sample.
  • Molecular formula C21H22O8; molecular weight 402.39.
Catalog No. 50-002-96349 Supplier Medchemexpress LLC Supplier No. HYN015510MG
Nobiletin is a poly-methoxylated flavone derived from citrus peel. It acts as a retinoid acid receptor-related orphan receptor (ROR) agonist, modulates reactive oxygen species (ROS), and has reported antioxidant, anti-inflammatory, neuroprotective, and anti-cancer activities. Supplied as a powdered research compound; follow storage guidelines in the product datasheet.
  • Poly-methoxylated flavone with ROR agonist activity.
  • Modulates ROS and influences apoptosis and autophagy pathways.
  • Suitable for in vitro and preclinical research applications.
  • Supplied as a powder; available as a 10 MG research sample.
  • Molecular formula C21H22O8; molecular weight 402.39.