Learn More
Selleck Chemical LLC Nobiletin 50mg 478-01-3 Hexamethoxyflavone
SDP
Catalog No. 505361
Encompass_Preferred
Nobiletin (Hexamethoxyflavone), a citrus flavonoid isolated from citrus peels like in tangerine, which has anti-inflammatory and anti-tumor activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-536-1 Supplier Selleck Chemical LLC Supplier No. S233350MG
Nobiletin (Hexamethoxyflavone), a citrus flavonoid isolated from citrus peels like in tangerine, which has anti-inflammatory and anti-tumor activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.