Learn More
GENERAL BIOPHYSICS LLC Universal Noble Gas Analyzer Model A
Catalog No. NC3361694
Encompass
Thermoconductivity Gas Analyzer for Noble Gases with Onboard PumpApplication: Calibrated for noble gases (Xe, Ar, Ne, Kr, He) with onboard pump to take gas samples.Features: High accuracy for all noble gases.
Catalog No. NC3361694 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAUNIVA
Add to Cart
Add to Cart
Request a Quote
Thermoconductivity Gas Analyzer for Noble Gases with Onboard PumpApplication: Calibrated for noble gases (Xe, Ar, Ne, Kr, He) with onboard pump to take gas samples.Features: High accuracy for all noble gases.