Learn More
GENERAL BIOPHYSICS LLC Universal Noble Gas Analyzer Model B
Supplier:  GENERAL BIOPHYSICS LLC GAUNIVB
Encompass
Thermoconductivity Gas Analyzer for Noble Gases (In-line Monitoring)Application: Real-time, in-line monitoring of noble gases (Xe, Ar, Ne, Kr, He).
Catalog No. NC3361697
Add to Cart
Request a Quote