Learn More
Aviva Systems Biology Corp NOBOX Antibody - N-terminal region (ARP40110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOBOX. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human NOBOX. Alternate names: OG2, OG-2, OG2X, POF5, TCAG_12042ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57294