Learn More
Enzo Life Sciences NOC-12 (5mg). CAS: 146724-89-2
SDP
Supplier:  Enzo Life Sciences ALX430019M005
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time of 0.1mM NOC-12 is 100 min. at 37°C in 0.1M PBS, pH 7.4. Alternative name: 1-Hydroxy-2-oxo-3-(N-ethyl-2-aminoethyl)-3-ethyl-1-triazene. Purity: ≥97% (HPLC). Solubility: Soluble in 0.1M NaOH (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9151