Learn More
Medchemexpress LLC HY-136278 10mg Medchemexpress, NOC 18 CAS:146724-94-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13627810MG
Encompass_Preferred
Medchemexpress, HY-136278 10mg NOC 18 CAS:146724-94-9 NOC 18 is a nitric oxide donor and activates an inward current in cultured rat cerebellar granules cells. NOC 18 increases cGMP production in cultured vascular smooth muscle cells. NOC 18 reduces contractility of cardiac muscle preparations in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1430