Learn More
Aviva Systems Biology Corp NOC4L Antibody - C-terminal region (ARP41005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOC4L. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NOC4L. Alternate names: NOC4, NET49, UTP19ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-57694