Learn More
AdipoGen Nocardamine (1 mg)
SDP
Supplier:  AdipoGen AGCN20150M001
Encompass_Preferred
Chemical. CAS: 26605-16-3. Formula: C27H48N6O9. MW: 600.7. Isolated from Streptomyces sp. K04-0144. Antibiotic. Anti-mycobacterial. Siderophore (iron (Fe) chelating compound). Inducer of morphological changes in insect cells. Antioxidant. Anticancer compound.
Catalog No. 50-205-3531