Learn More
ATCC Nocardia nova; Strain UT_1954_0606
Catalog No. 502383458
Encompass
Nocardia nova strain UT_1954_0606 was isolated in 1988 from a human in Texas. It has applications as a CLSI quality control organism for broth microdilution susceptibility testing of Nocardia species.
Catalog No. 50-238-3458 Supplier ATCC Supplier No. BAA2227
Nocardia nova strain UT_1954_0606 was isolated in 1988 from a human in Texas. It has applications as a CLSI quality control organism for broth microdilution susceptibility testing of Nocardia species.