Learn More
Sigma Aldrich Fine Chemicals Biosciences Alnochromix, 1130 g, 10/pk
Catalog No. NC0826175
Encompass
Each package, when mixed with sulfuric acid, makes 5 pt of cleaning solution. Metal-free, non-polluting, EPA recommended, non-carcinogenic. Box of 10 packs (10×3.1 oz packs).
Catalog No. NC0826175 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 32869310PAK
Each package, when mixed with sulfuric acid, makes 5 pt of cleaning solution. Metal-free, non-polluting, EPA recommended, non-carcinogenic. Box of 10 packs (10×3.1 oz packs).