Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Godax Labs™ Nochromix™ Reagent

Catalog No. 0434520 Shop All Alconox Products
Change view
Click to view available options
Volume (English):
3 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
04-345-20 3 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-345-20 Supplier Godax Labs™ Supplier No. 19 010/DEL

missing translation for 'validMainframeMsg'

Chemically cleans glassware

  • Inorganic oxidizer
  • Valuable for trace analysis, enzymology, and tissue culture work
  • Free-rinsing
  • Contains no metal ions
  • Each hermetically sealed 3 oz. (89mL) packet should be mixed with 1 gal. (3.8L) of concentrated sulfuric acid

Specifications

Form Crystalline
Appearance White
Container Type Hermetically sealed packet
For Use With (Application) Chemically cleans Glassware for trace analysis, enzymology and Tissue Culture work
Volume (English) 3 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.