Learn More
Bachem Nociceptin Trifluoroacetate
SDP
Supplier:  Bachem 4031112.0001
Encompass_Preferred
Nociceptin Trifluoroacetate, H-Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-Leu-Ala-Asn-Gln-OH, 170713-75-4, C79H129N27O22, Mr 1809.06, 1mg. Orphanin FQ. Nociceptin is an endogenous agonist of the opioid receptor-like ORL1 receptor.
Catalog No. 50-259-689