Learn More
CPC Scientific H-Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OPIO011A
Encompass
This agonist of nociceptin (H-3036) is the smallest peptide maintaining the same efficacy and potency as the natural peptide. The amidation protects it from degradation by proteases.ONE-LETTER SEQUENCE: FGGFTGARKSARK-NH2MOLECULAR FORMULA: C61H100N22O15MOLECULAR WEIGHT:1381.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [178064-02-3], RESEARCH AREA: NeuropeptidesREFERENCES: G.Calo et al., Can. J. Physiol. Pharmacol., 75, 713 (1997); R.Guerrini et al., J. Med. Chem., 40, 1789 (1997)
Catalog No. 50-210-9984