Learn More
CPC Scientific H-Thr-Glu-Pro-Gly-Leu-Glu-Glu-Val-Gly-Glu-Ile-Glu-Gln-Lys-Gln-Leu-Gln-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP120A
Encompass
ONE-LETTER SEQUENCE: TEPGLEEVGEIEQKQLQMOLECULAR FORMULA: 82H135N21O32MOLECULAR WEIGHT:1927.10STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [208253-85-4], SYNONYMS: Nocistatin (bovine)RESEARCH AREA: NeuropeptidesREFERENCES: E. Okuda-Ashitaka et al., Nature, 392, 286 (1998)
Catalog No. 50-210-9954