Learn More
Brew Plate™ Nocive Brewers Bacteria Agar - 500G

Description
Uses:
- Culturing microorganisms
Specifications
Specifications
| Content And Storage | 2-30° C |
| Product Type | Agar |
| For Use With (Application) | Cultivating yeast and bacteria found in brewing |
| Quantity | 1 EA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.