Learn More
Enzo Life Sciences Nocodazole, (10mg), CAS Number: 31430-18-9
SDP
Supplier:  Enzo Life Sciences BMLT1010010
Encompass_Preferred
Microtubule inhibitor. Synthetic reversible microtubule inhibitor. Alternative Name: Methyl[5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl]carbamate. R 17934. Formula: C14H11N3O3S. MW: 301.3. CAS Number: 31430-18-9. Purity: ≥98% (HPLC). Appearance: Solid. White, off-white, orange and possibly even a brown/pink color. Solubility: Soluble in DMSO (10mg/ml) or dimethyl formamide; almost insoluble in water. Use/Stability: Stable for at least 1 year after receipt when stored at +20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: Ambient
Catalog No. NC9038247